Assay Details
Gene Symbol: KCNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206413.3 | NP_001193342.1 | 1 - 2 | 4712 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
