Assay Details
| Gene Symbol: CBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206415.1 | NP_001193344.1 | 3 - 4 | 555 | 66 | |
| GenBank mRNA | DV886299.1 | - | 3 - 4 | 491 | 66 | |
More Information
Set Membership: |
|

CBX1
Genomic Map