Assay Details
| Gene Symbol: BIVM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206453.2 | NP_001193382.2 | 3 - 4 | 1103 | 92 | |
| GenBank mRNA | DV823208.1 | - | 2 - 3 | 175 | 92 | |
| EE907980.1 | - | 1 - 2 | 106 | 92 | ||
More Information
Set Membership: |
|

BIVM
Genomic Map