Assay Details
| Gene Symbol: PML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206503.1 | NP_001193432.1 | 7 - 8 | 1770 | 64 | |
| GenBank mRNA | BC149786.1 | - | 7 - 8 | 1731 | 64 | |
More Information
Set Membership: |
|

PML
Genomic Map