Assay Details
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206509.1 | NP_001193438.1 | 5 - 6 | 715 | 75 | |
GenBank mRNA | EV658200.1 | - | 5 - 6 | 717 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
