Assay Details
| Gene Symbol: HMGCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206578.1 | NP_001193507.1 | 10 - 11 | 1691 | 80 | |
| GenBank mRNA | AY581197.1 | - | 5 - 6 | 666 | 80 | |
| BC102850.1 | - | 10 - 11 | 1601 | 80 | ||
| EV627373.1 | - | 2 - 3 | 243 | 80 | ||
More Information
Set Membership: |
|

HMGCS1
Genomic Map