Assay Details
Gene Symbol: CHST11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192668.3 | NP_001179597.1 | 1 - 2 | 244 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
