Assay Details
| Gene Symbol: IGSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192700.2 | NP_001179629.2 | 3 - 4 | 833 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IGSF3| Gene Symbol: IGSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192700.2 | NP_001179629.2 | 3 - 4 | 833 | 52 | |
Set Membership: |
|