Assay Details
Gene Symbol: TUBA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166505.1 | NP_001159977.1 | 1 - 2 | 345 | 52 | |
GenBank mRNA | EH175948.1 | - | 1 - 2 | 345 | 52 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001166505.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
