Assay Details
Gene Symbol: PAK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206594.2 | NP_001193523.2 | 7 - 8 | 2068 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
