Assay Details
| Gene Symbol: GSTT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206623.1 | NP_001193552.1 | 2 - 3 | 574 | 59 | |
| GenBank mRNA | CR848913.2 | - | 2 - 3 | 298 | 59 | |
More Information
Set Membership: |
|

GSTT4
Genomic Map