Assay Details
| Gene Symbol: COA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206642.1 | NP_001193571.1 | 1 - 2 | 216 | 87 | |
| GenBank mRNA | EV628191.1 | - | 1 - 2 | 222 | 87 | |
| EV715580.1 | - | 2 - 3 | 521 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206642.1 transcripts.
Set Membership: |
|

COA6
Genomic Map