Assay Details
| Gene Symbol: ENPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206724.1 | NP_001193653.1 | 3 - 4 | 1066 | 102 | |
| GenBank mRNA | DV884416.1 | - | 1 - 2 | 108 | 102 | |
| DV913973.1 | - | 2 - 3 | 614 | 102 | ||
More Information
Set Membership: |
|

ENPP5
Genomic Map