Assay Details
Gene Symbol: NCR3LG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206792.1 | NP_001193721.1 | 3 - 4 | 1020 | 89 | |
GenBank mRNA | EH166075.1 | - | 2 - 3 | 421 | 89 | |
MH237865.1 | - | 3 - 4 | 924 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
