Assay Details
| Gene Symbol: PDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166527.1 | NP_001159999.1 | 1 - 2 | 56 | 104 | |
| GenBank mRNA | M33529.1 | - | 1 - 2 | 37 | 104 | |
More Information
Set Membership: |
|

PDC
Genomic Map