Assay Details
| Gene Symbol: PDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166527.1 | NP_001159999.1 | 2 - 3 | 216 | 119 | |
| GenBank mRNA | M33529.1 | - | 2 - 3 | 197 | 119 | |
More Information
Set Membership: |
|

PDC
Genomic Map