Assay Details
| Gene Symbol: LAMTOR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166607.1 | NP_001160079.1 | 1 - 2 | 82 | 88 | |
| GenBank mRNA | EH136813.1 | - | 1 - 2 | 83 | 88 | |
| EH144437.1 | - | 1 - 2 | 53 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001166607.1 transcripts.
Set Membership: |
|

LAMTOR4
Genomic Map