Assay Details
| Gene Symbol: LAMTOR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166607.1 | NP_001160079.1 | 2 - 3 | 156 | 77 | |
| GenBank mRNA | BC111612.1 | - | 98 | 77 | ||
| EH136813.1 | - | 2 - 3 | 157 | 77 | ||
| EH144437.1 | - | 2 - 3 | 127 | 77 | ||
More Information
Set Membership: |
|

LAMTOR4
Genomic Map