Assay Details
| Gene Symbol: MGARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166611.1 | NP_001160083.1 | 1 - 2 | 156 | 98 | |
| GenBank mRNA | BC148137.1 | - | 1 - 2 | 105 | 98 | |
| EV659412.1 | - | 1 - 2 | 165 | 98 | ||
More Information
Set Membership: |
|

MGARP
Genomic Map