Assay Details
| Gene Symbol: PEG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172518.2 | NP_001165989.2 | 1 - 2 | 211 | 79 | |
| NM_001172519.2 | NP_001165990.2 | 1 - 2 | 211 | 79 | ||
| GenBank mRNA | CN438370.1 | - | 1 - 2 | 188 | 79 | |
| EV615229.1 | - | 1 - 2 | 217 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001172518.2, NM_001172519.2 transcripts.
Set Membership: |
|

PEG10
Genomic Map