Assay Details
| Gene Symbol: DDX3Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172595.1 | NP_001166066.1 | 10 - 11 | 1612 | 109 | |
| GenBank mRNA | FJ659845.1 | - | 10 - 11 | 1503 | 109 | |
| GQ259590.1 | - | 1 | 1612 | 109 | ||
| GQ259591.1 | - | 10 - 11 | 1612 | 109 | ||
More Information
Set Membership: |
|

DDX3Y
Genomic Map