Assay Details
| Gene Symbol: DDX3Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172595.1 | NP_001166066.1 | 161 | 99 | ||
| GenBank mRNA | FJ659845.1 | - | 52 | 99 | ||
| GQ259590.1 | - | 161 | 99 | |||
| GQ259591.1 | - | 161 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX3Y| Gene Symbol: DDX3Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172595.1 | NP_001166066.1 | 161 | 99 | ||
| GenBank mRNA | FJ659845.1 | - | 52 | 99 | ||
| GQ259590.1 | - | 161 | 99 | |||
| GQ259591.1 | - | 161 | 99 | |||