Assay Details
| Gene Symbol: OAZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172708.1 | NP_001166179.1 | 2 - 3 | 335 | 60 | |
| GenBank mRNA | DT853300.1 | - | 2 - 3 | 335 | 60 | |
| EH124932.1 | - | 2 - 3 | 255 | 60 | ||
More Information
Set Membership: |
|

OAZ3
Genomic Map