Assay Details
Gene Symbol: LYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177740.2 | NP_001171211.2 | 8 - 9 | 1197 | 58 | |
GenBank mRNA | AB553580.1 | - | 8 - 9 | 1197 | 58 | |
AB562971.1 | - | 8 - 9 | 1197 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
