Assay Details
| Gene Symbol: NUDT19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191214.1 | NP_001178143.1 | 2 - 3 | 1050 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUDT19| Gene Symbol: NUDT19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191214.1 | NP_001178143.1 | 2 - 3 | 1050 | 122 | |
Set Membership: |
|