Assay Details
| Gene Symbol: ELK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191236.1 | NP_001178165.1 | 2 - 3 | 331 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ELK1| Gene Symbol: ELK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191236.1 | NP_001178165.1 | 2 - 3 | 331 | 60 | |
Set Membership: |
|