Assay Details
Gene Symbol: ZACN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191252.3 | NP_001178181.1 | 3 - 4 | 269 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
