Assay Details
| Gene Symbol: GBA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191277.2 | NP_001178206.2 | 1 - 2 | 85 | 61 | |
| GenBank mRNA | GH317543.1 | - | 1 - 2 | 85 | 61 | |
More Information
Set Membership: |
|

GBA3
Genomic Map