Assay Details
| Gene Symbol: AJAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206815.1 | NP_001193744.1 | 3 - 4 | 1040 | 82 | |
| GenBank mRNA | EV704037.1 | - | 3 - 4 | 417 | 82 | |
More Information
Set Membership: |
|

AJAP1
Genomic Map