Assay Details
| Gene Symbol: FGF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206832.2 | NP_001193761.1 | 4 - 5 | 628 | 64 | |
| GenBank mRNA | DT887517.1 | - | 1 - 2 | 279 | 64 | |
More Information
Set Membership: |
|

FGF14
Genomic Map