Assay Details
| Gene Symbol: TMPRSS11BNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206871.1 | NP_001193800.1 | 2 - 3 | 211 | 100 | |
| GenBank mRNA | EV690549.1 | - | 2 - 3 | 212 | 100 | |
More Information
Set Membership: |
|

TMPRSS11BNL
Genomic Map