Assay Details
| Gene Symbol: TMPRSS11BNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206871.1 | NP_001193800.1 | 3 - 4 | 333 | 127 | |
| GenBank mRNA | EV690549.1 | - | 3 - 4 | 334 | 127 | |
More Information
Set Membership: |
|

TMPRSS11BNL
Genomic Map