Assay Details
| Gene Symbol: TMPRSS11BNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206871.1 | NP_001193800.1 | 4 - 5 | 395 | 102 | |
| GenBank mRNA | EV690549.1 | - | 4 - 5 | 396 | 102 | |
More Information
Set Membership: |
|

TMPRSS11BNL
Genomic Map