Assay Details
Gene Symbol: TMPRSS11BNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206871.1 | NP_001193800.1 | 6 - 7 | 599 | 99 | |
GenBank mRNA | EV690549.1 | - | 6 - 7 | 600 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
