Assay Details
| Gene Symbol: TMPRSS11BNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206871.1 | NP_001193800.1 | 7 - 8 | 772 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS11BNL| Gene Symbol: TMPRSS11BNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206871.1 | NP_001193800.1 | 7 - 8 | 772 | 83 | |
Set Membership: |
|