Assay Details
Gene Symbol: MNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206899.2 | NP_001193828.1 | 1 - 2 | 71 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
