Assay Details
| Gene Symbol: MTNR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206907.2 | NP_001193836.2 | 1 - 2 | 224 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTNR1B| Gene Symbol: MTNR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206907.2 | NP_001193836.2 | 1 - 2 | 224 | 53 | |
Set Membership: |
|