Assay Details
Gene Symbol: PSMC3IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244194.1 | NP_001231123.1 | 3 - 4 | 246 | 80 | |
GenBank mRNA | AW425616.1 | - | 3 - 4 | 247 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
