Assay Details
| Gene Symbol: NOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191340.1 | NP_001178269.1 | 7 - 8 | 798 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOX1| Gene Symbol: NOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191340.1 | NP_001178269.1 | 7 - 8 | 798 | 61 | |
Set Membership: |
|