Assay Details
Gene Symbol: TFPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244204.1 | NP_001231133.1 | 4 - 5 | 513 | 82 | |
GenBank mRNA | BC116160.1 | - | 4 - 5 | 502 | 82 | |
EE251296.1 | - | 4 - 5 | 514 | 82 | ||
EV744049.1 | - | 4 - 5 | 602 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
