Assay Details
| Gene Symbol: MTERF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244226.1 | NP_001231155.1 | 3 - 4 | 728 | 61 | |
| GenBank mRNA | BC104566.1 | - | 2 - 3 | 666 | 61 | |
More Information
Set Membership: |
|

MTERF4
Genomic Map