Assay Details
| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173959.4 | NP_776384.3 | 5 - 6 | 1024 | 89 | |
| GenBank mRNA | AB075020.1 | - | 5 - 6 | 1267 | 89 | |
| AY241933.1 | - | 5 - 6 | 1024 | 89 | ||
| BC112700.1 | - | 5 - 6 | 1274 | 89 | ||
More Information
Set Membership: |
|

SCD
Genomic Map