Assay Details
| Gene Symbol: AVPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174246.2 | NP_776671.1 | 2 - 3 | 101 | 100 | |
| GenBank mRNA | X83741.1 | - | 101 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174246.2, X83741.1 transcripts.
Set Membership: |
|

AVPR2
Genomic Map