Assay Details
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174660.2 | NP_777085.1 | 2 - 3 | 658 | 76 | |
GenBank mRNA | BC150034.1 | - | 2 - 3 | 744 | 76 | |
BT029910.1 | - | 2 - 3 | 675 | 76 | ||
M24103.1 | - | 2 - 3 | 659 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
