Assay Details
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174660.2 | NP_777085.1 | 3 - 4 | 802 | 64 | |
GenBank mRNA | BC150034.1 | - | 3 - 4 | 888 | 64 | |
BT029910.1 | - | 3 - 4 | 819 | 64 | ||
M24103.1 | - | 3 - 4 | 803 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
