Assay Details
| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191482.1 | NP_001178411.1 | 3 - 4 | 281 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191482.1 transcripts.
Set Membership: |
|

ZNF248
Genomic Map