Assay Details
| Gene Symbol: GRHPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192037.3 | NP_001178966.1 | 1 - 2 | 110 | 89 | |
| GenBank mRNA | BC120229.1 | - | 1 - 2 | 115 | 89 | |
More Information
Set Membership: |
|

GRHPR
Genomic Map