Assay Details
| Gene Symbol: ALOX12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192038.2 | NP_001178967.1 | 12 - 13 | 1650 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALOX12B| Gene Symbol: ALOX12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192038.2 | NP_001178967.1 | 12 - 13 | 1650 | 64 | |
Set Membership: |
|