Assay Details
| Gene Symbol: FXYD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192063.2 | NP_001178992.1 | 2 - 3 | 316 | 65 | |
| GenBank mRNA | BC113304.1 | - | 2 - 3 | 135 | 65 | |
More Information
Set Membership: |
|

FXYD5
Genomic Map