Assay Details
| Gene Symbol: PLA2G2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192086.1 | NP_001179015.1 | 3 - 4 | 289 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G2E| Gene Symbol: PLA2G2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192086.1 | NP_001179015.1 | 3 - 4 | 289 | 93 | |
Set Membership: |
|